- Loss of CPSF2 expression is associated with increased thyroid cancer cellular invasion and cancer stem cell population, and more aggressive disease.
Loss of CPSF2 expression is associated with increased thyroid cancer cellular invasion and cancer stem cell population, and more aggressive disease.
Identification of molecular factors that promote thyroid cancer progression have important clinical implications for therapy and prognostication in patients with papillary thyroid cancer (PTC). The aim of this study was to validate and determine the function of dysregulated genes that were associated increased mortality in patients with PTC. Experiemental Design: We selected the cleavage and polyadenylation specificity factor subunit 2 (CPSF2) gene from the top 5 significantly dysregulated genes associated with PTC-associated mortality from our previous study. We used 86 PTC samples enriched for aggressive disease (recurrence and mortality) by quantitative RT-PCR (qRT-PCR). In vitro functional studies of the validated gene were performed. Decreased CPSF2 gene expression was associated with shorter disease-free survival (P = .03), large tumor size (T3 and T4) (P = .03), tumor recurrence (P < .01), and mortality (P < .01), independent of BRAF V600E mutation status. CPSF2 knockdown increased cellular invasion by 1.8- to 3.2-fold (P < .01) and increased markers of thyroid cancer stem cells (CD44 and CD133 expression). Immunohistochemistry showed an inverse correlation between CD44 protein expression in PTC samples and CPSF2 expression. Decreased CPSF2 expression is associated with increased cellular invasion and cancer stem cell population, and more aggressive disease in PTC.