Merck
Hyperbranched lead selenide nanowire networks.
Nano letters (2007-03-14)
Jia Zhu, Hailin Peng, Candace K Chan, Konrad Jarausch, Xiao Feng Zhang, Yi Cui
ABSTRACT
Lead chalcogenide nanostructures are good potential candidates for applications in multiexciton solar cells, infrared photodetectors, and electroluminescence devices. Here we report the synthesis and electrical measurements of hyperbranched PbSe nanowire networks. Hyperbranched PbSe nanowire networks are synthesized via a vapor-liquid-solid (VLS) mechanism. The branching is induced by continuously feeding the PbSe reactant with the vapor of a low-melting-point metal catalyst including In, Ga, and Bi. The branches show very regular orientation relationships: either perpendicular or parallel to each other. The diameter of the individual NWs depends on the size of the catalyst droplets, which can be controlled by the catalyst vapor pressure. Significantly, the hyperbranched networks can be grown epitaxially on NaCl, a low-cost substrate for future device array applications. Electrical measurements across branched NWs show the evolution of charge carrier transport with distance and degree of branching.