Merck
  • HIF-1 stabilization exerts anticancer effects in breast cancer cells in vitro and in vivo.
HIF-1 stabilization exerts anticancer effects in breast cancer cells in vitro and in vivo.
Biochemical pharmacology (2020-03-25)
Neli Kachamakova-Trojanowska, Paulina Podkalicka, Tomasz Bogacz, Szymon Barwacz, Alicja Józkowicz, Józef Dulak, Agnieszka Łoboda
ABSTRACT
Tumor hypoxia and high activity of hypoxia-inducible factor-1 (HIF-1) correlate with adverse disease outcomes, malignancy, resistance to therapy and metastasis. Nonetheless, recent studies indicate that under certain circumstances, HIF-1 stabilization may exert protective effects and even decrease tumor cell aggressiveness. This study aimed to characterize the potential anticancer effect of molidustat (BAY 85-3934), the prolyl hydroxylase (PHD) inhibitor and HIF-1 stabilizator. We confirmed that molidustat stabilizes HIF-1α and induces the expression of vascular endothelial growth factor (VEGF) in MDA-MB-231 breast cancer cells, to a similar or even greater extent than hypoxia. Interestingly, decreased cell survival and colony formation capabilities, together with S/G2 cell cycle arrest, were observed after treatment with PHD inhibitor. Importantly, molidustat enhanced the effectiveness of the chemotherapeutic drug, gemcitabine, on cancer cells. Finally, the xenograft model revealed decreased tumor growth in vivo after molidustat treatment. Both in vitro and in vivo analysis showed no differences in the angiogenic potential of endothelial cells treated with tumor-conditioned media or vascularization of the MDA-MB-231 xenografts, respectively. In summary, molidustat treatment exhibits an inhibitory effect on breast cancer cell survival, self-renewal capacity and potentiates the efficacy of chemotherapeutic gemcitabine.