Bis(2-ethylhexyl) phthalate
Synonym(s):
DEHP, Phthalic acid bis(2-ethylhexyl ester), ‘Dioctyl’ phthalate, Bis(2-ethylhexyl) phthalate
Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing