Proteomix® WAX-NP10 Weak Anion Exchange Column
Products
Product Category
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing