Skip to Content
SG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium nitrite
Linear Formula:
NaNO
2
CAS Number:
7632-00-0
Molecular Weight:
69.00
EC Number:
231-555-9
Products
Product Category
salts (7)
Brand
Sigma-Aldrich (7)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (3)
Anhydrous (2)
Puriss (2)
Puriss P.A. (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
237213
Sodium nitrite,
ACS reagent, ≥97.0%
Expand
View Pricing
S2252
Sodium nitrite,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
563218
Sodium nitrite,
99.999% trace metals basis
Expand
View Pricing
31443
Sodium nitrite,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Expand
View Pricing
901903
Sodium nitrite,
anhydrous, Redi-Dri
™
,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
799416
Sodium nitrite,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥97%
Expand
View Pricing
13447
Sodium nitrite,
puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 250, 99-100.5% (calc. to the dried substance)
Expand
View Pricing