Merck
  • A versatile approach for the asymmetric syntheses of (1R,9aR)-epiquinamide and (1R,9aR)-homopumiliotoxin 223G.
A versatile approach for the asymmetric syntheses of (1R,9aR)-epiquinamide and (1R,9aR)-homopumiliotoxin 223G.
Organic letters (2006-03-28)
Pei-Qiang Huang, Zheng-Qing Guo, Yuan-Ping Ruan
ABSTRACT
[reaction: see text] Using 5b as a common intermediate, the first asymmetric synthesis of (-)-epiquinamide (4) and a formal asymmetric synthesis of (-)-homopumiliotoxin 223G (2) is described. A key feature of our approach is the flexible introduction of a functionalized C(4) side chain to (S)-3-benzyloxyglutarimide 7 in a regio- and diastereoselective manner. Utilization of a tandem Swern oxidation-Grignard addition strategy efficiently prevented racemization. An unexpected NaN(3)-promoted methanesulfonic acid elimination yielded 17, a reaction which could be useful for the syntheses of 8-dehydrodesmethylpumiliotoxins such as alkaloid 235C (3).
Sigma-Aldrich
Glutarimide, 98%