Merck
  • Synthesis and Bioactivity Evaluation of a Novel 1,2,4-Oxadiazole Derivative in vitro and in 3×Tg Mice.
Synthesis and Bioactivity Evaluation of a Novel 1,2,4-Oxadiazole Derivative in vitro and in 3×Tg Mice.
Drug design, development and therapy (2022-10-04)
Zhuohui Luo, Yongcheng Wang, Shuo Pang, Shan Gao, Ning Liu, Xiang Gao, Li Zhang, Xiaolong Qi, Yajun Yang, Lianfeng Zhang
ABSTRACT
Alzheimer's disease (AD) is the most common neurodegenerative disease whose patients suffered from cognitive impairments. In our study, a novel 1,2,4-Oxadiazole derivative wyc-7-20 was synthesized, which showed low cytotoxicity and potent neuroprotective effect at the cellular level. Improved cognitive impairments, β-amyloid (Aβ) clearance, and tau pathological phenotypes were detected in transgenic animal models after wyc-7-20 treatment. Reversed expressions in AD-related genes were also detected. The results demonstrated wyc-7-20 was potent in AD therapy. The pathological complexity of AD increased difficulties in medical research. To explore a new potential medical treatment for AD, a novel 1,2,4-Oxadiazole derivative (wyc-7-20) was designed, synthesized to explore the application in this study. Human neuroblastoma (SH-SY5Y) cells and human hepatocellular carcinoma (HepG2) cells were used to detect median lethal dose (LD50). H2O2 and Aβ1-42 oligomers (AβOs) were respectively, added into SH-SY5Y cells to detect anti-ROS (reactive oxygen species) and anti-AβOs effects of wyc-7-20. 3×Tg mice were administered with wyc-7-20, and then Y maze test and Morris water maze (MWM) test were applied to detect cognitive improvements. Brain tissue samples were subsequently collected and analyzed using different techniques. wyc-7-20 showed low cytotoxicity and potent neuroprotective effect at the cellular level. Improved cognitive impairments, Aβ clearance, and tau pathological phenotypes were detected in transgenic animal models after wyc-7-20 treatment. Reversed expressions in AD-related genes were also detected. wyc-7-20 was potent in AD therapy.
Sigma-Aldrich
Anti-Tau Mouse mAb (TAU-5), liquid, clone TAU-5, Calbiochem®