Merck
Spinal TASK-1 and TASK-3 modulate inflammatory and neuropathic pain.
European journal of pharmacology (2019-09-01)
Guadalupe García, Roxana Noriega-Navarro, Vladimir A Martínez-Rojas, Erika J Gutiérrez-Lara, Norma Oviedo, Janet Murbartián
ABSTRACT
This study assessed the participation of spinal TWIK-related acid-sensitive K+ channels 1 and 3 (TASK-1 and TASK-3) in inflammatory (formalin test) and neuropathic (spinal nerve ligation, SNL) pain in rats. Intrathecal pre-treatment (-10 min) with the TASK-1 blocker ML365 or TASK-3 blocker PK-THPP, but not vehicle, enhanced in a dose-dependent manner 1% formalin-induced acute and long-lasting secondary mechanical allodynia and mechanical hyperalgesia in rats. In contrast, intrathecal pre-treatment with terbinafine, an activator of TASK-3, reduced formalin-induced flinching and allodynia/hyperalgesia. Both blockers and terbinafine had similar effects on female and male rats. In addition, intrathecal injection of ML365 or PK-THPP blocked the terbinafine-induced antiallodynic effect in neuropathic rats, but they did not modify baseline withdrawal threshold in naïve or sham-operated rats. TASK-1 and TASK-3 mRNA and protein were expressed in L4 and L5 dorsal root ganglia (DRG) and dorsal and ventral spinal cord of naïve animals. Interestingly, formalin injection increased TASK-1 expression in ipsilateral L5 DRG, but not in the spinal cord. Moreover, formalin injection transiently enhanced TASK-3 expression in ipsilateral L5 DRG and dorsal spinal cord. In contrast, SNL down-regulated TASK-3 expression in the ipsilateral L4 and L5 DRG but not in dorsal or ventral spinal cord, while SNL did not modify TASK-1 expression at any tissue. The pharmacological and molecular results suggest that TASK-1 and TASK-3 have a relevant antinociceptive role in inflammatory and neuropathic pain.