Merck
  • The PtdIns3P phosphatase myotubularin is a cytoplasmic protein that also localizes to Rac1-inducible plasma membrane ruffles.
The PtdIns3P phosphatase myotubularin is a cytoplasmic protein that also localizes to Rac1-inducible plasma membrane ruffles.
Journal of cell science (2002-07-16)
Jocelyn Laporte, Francois Blondeau, Anne Gansmuller, Yves Lutz, Jean-Luc Vonesch, Jean-Louis Mandel
ABSTRACT
Myotubularin, the phosphatase mutated in X-linked myotubular myopathy, was shown to dephosphorylate phosphatidylinositol 3-monophosphate (PtdIns3P) and was also reported to interact with nuclear transcriptional regulators from the trithorax family. We have characterized a panel of specific antibodies and investigated the subcellular localization of myotubularin. Myotubularin is not detected in the nucleus, and localizes mostly as a dense cytoplasmic network. Overexpression of myotubularin does not detectably affect vesicle trafficking in the mammalian cells investigated, in contrast to previous observations in yeast models. Both mutation of a key aspartate residue of myotubularin and dominant activation of Rac1 GTPase lead to the recruitment of myotubularin to specific plasma membrane domains. Localization to Rac1-induced ruffles is dependent on the presence of a domain highly conserved in the myotubularin family (that we named RID). We thus propose that myotubularin may dephosphorylate a subpool of PtdIns3P (or another related substrate) at the plasma membrane.