![Gluconapin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/168/909/1cd08bb3-d8e0-4f5f-959c-6d3c532f9cf4/640/1cd08bb3-d8e0-4f5f-959c-6d3c532f9cf4.png)
Gluconapin potassium salt
![Gluconapin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/168/909/1cd08bb3-d8e0-4f5f-959c-6d3c532f9cf4/640/1cd08bb3-d8e0-4f5f-959c-6d3c532f9cf4.png)
Synonym(s):
3-Butenylglucosinolate potassium salt
Empirical Formula (Hill Notation):
C11H18KNO9S2
CAS Number:
Molecular Weight:
411.49
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing