Clarithromycin
Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing