(S)-(+)-Camptothecin
Synonym(s):
Camptothecin, Camptotheca acuminata
Empirical Formula (Hill Notation):
C20H16N2O4
CAS Number:
Molecular Weight:
348.35
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing