Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9172-98

CY®3 Amidite 100mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.32

form

solid

manufacturer/tradename

Cytiva

color

Red

solubility

soluble (Soluble in cold water and hot water)

General description

For incorporation of Cy3 fluorescent dyes at any position in an oligonucleotide during synthesis.

Linkage of Cy dyes allows labeled DNA to hybridize to other DNA.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Other Notes

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use

Legal Information

Cy is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service