HPLC Analysis of Toluene, Uracil and Cytosine on Chromolith® Performance Diol 100-4.6mm
CONDITIONS
column
Chromolith® Performance DIOL, 100 x 4.6 mm (1.53172.0001)
mobile phase
[A] acetonitrile; [B] 5mM Ammoniumacetate buffer pH=6.8; (80:20, A:B)
flow rate
0.5 mL/min
pressure
87 psi (6 Bar)
column temp.
23 °C
detector
254 nm
injection
10 μL
sample
Toluene 0,67mg/ml, Uracil 0,04mg/ml, Cytosine 0,06mg/ml (Diluent: Mobile phase)
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany