Merck
  • Base-Modified Nucleic Acids as a Powerful Tool for Synthetic Biology and Biotechnology.
Base-Modified Nucleic Acids as a Powerful Tool for Synthetic Biology and Biotechnology.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-05-18)
Elena Eremeeva, Michail Abramov, Lia Margamuljana, Piet Herdewijn
ABSTRACT
The ability of various nucleoside triphosphate analogues of deoxyguanosine and deoxycytidine with 7-deazadeoxyadenosine (A