Merck
  • TWEAK can induce pro-inflammatory cytokines and matrix metalloproteinase-9 in macrophages.
TWEAK can induce pro-inflammatory cytokines and matrix metalloproteinase-9 in macrophages.
Circulation journal : official journal of the Japanese Circulation Society (2004-04-02)
Se-Hwa Kim, Yoon-Joong Kang, Won-Jung Kim, Dong-Kyun Woo, Yoon Lee, Dong-Ik Kim, Yong Bok Park, Byoung S Kwon, Jeong-Euy Park, Won-Ha Lee
ABSTRACT
The expression of TWEAK (TNFSF12) and TweakR/Fn14 was detected in regions rich in macrophage/foam cells in atherosclerotic plaques. The role of TWEAK in monocytes in relation to atherogenesis was investigated by analyzing the cellular events induced by TWEAK in a human macrophage-like cell line, THP-1. TWEAK induced various molecular mediators of atherogenesis, such as IL-6, MCP-1, IL-8 and MMP-9, and the induction was augmented by interferon-gamma. TWEAK-induced activation of MMP-9 was mediated by activation of NF-kappaB. These results suggest that TWEAK is involved in atherosclerosis by inducing pro-inflammatory cytokines and extracellular matrix degrading enzymes, which reduce plaque stability.