Merck
  • Use of site-directed antibodies to probe the topography of the alpha 2 subunit of voltage-gated Ca2+ channels.
Use of site-directed antibodies to probe the topography of the alpha 2 subunit of voltage-gated Ca2+ channels.
FEBS letters (1995-05-08)
K Brickley, V Campbell, N Berrow, R Leach, R I Norman, D Wray, A C Dolphin, S A Baldwin
ABSTRACT
Polyclonal antibodies were raised against peptides corresponding to residues 1-15, 469-483 and 933-951 of the rabbit skeletal muscle L-type calcium channel alpha 2/delta primary translation product, for use as topological probes. Immunocytochemical comparison of the abilities of the antibodies to bind to the alpha 2 and delta subunits in intact and detergent-permeabilised rat dorsal root ganglion cells enabled the membrane orientation of these regions to be established. The resultant data indicate that the regions containing residues 1-15 and 469-483 of the alpha 2 subunit, and residues 1-17 of the delta subunit, are exposed on the extracellular surface of the membrane, findings consistent with a model that proposes alpha 2 to be entirely extracellular.