Merck
  • Author Correction: Blunting neuroinflammation with resolvin D1 prevents early pathology in a rat model of Parkinson's disease.
Author Correction: Blunting neuroinflammation with resolvin D1 prevents early pathology in a rat model of Parkinson's disease.
Nature communications (2019-10-16)
Paraskevi Krashia, Alberto Cordella, Annalisa Nobili, Livia La Barbera, Mauro Federici, Alessandro Leuti, Federica Campanelli, Giuseppina Natale, Gioia Marino, Valeria Calabrese, Francescangelo Vedele, Veronica Ghiglieri, Barbara Picconi, Giulia Di Lazzaro, Tommaso Schirinzi, Giulia Sancesario, Nicolas Casadei, Olaf Riess, Sergio Bernardini, Antonio Pisani, Paolo Calabresi, Maria Teresa Viscomi, Charles Nicholas Serhan, Valerio Chiurchiù, Marcello D'Amelio, Nicola Biagio Mercuri
ABSTRACT
An amendment to this paper has been published and can be accessed via a link at the top of the paper.