Merck
  • Ubiquitin COOH-terminal hydrolase L1 deletion is associated with urinary α-klotho deficiency and perturbed phosphate homeostasis.
Ubiquitin COOH-terminal hydrolase L1 deletion is associated with urinary α-klotho deficiency and perturbed phosphate homeostasis.
American journal of physiology. Renal physiology (2018-04-19)
Naomi C Boisvert, Chet E Holterman, Alexey Gutsol, Josée Coulombe, Wanling Pan, R Todd Alexander, Douglas A Gray, Chris R Kennedy
ABSTRACT
Loss of ubiquitin COOH-terminal hydrolase L1 (UCHL1), a deubiquitinating enzyme required for neuronal function, led to hyperphosphatemia accompanied by phosphaturia in mice, while calcium homeostasis remained intact. We therefore investigated the mechanisms underlying the phosphate imbalance in Uchl1-/- mice. Interestingly, phosphaturia was not a result of lower renal brush border membrane sodium-phosphate cotransporter expression as sodium-phosphate cotransporter 2a and 2c expression levels was similar to wild-type levels. Plasma parathyroid hormone and fibroblast growth factor 23 levels were not different; however, fibroblast growth factor 23 mRNA levels were significantly increased in femur homogenates from Uchl1-/- mice. Full-length and soluble α-klotho levels were comparable in kidneys from wild-type and Uchl1-/- mice; however, soluble α-klotho was reduced in Uchl1-/- mice urine. Consistent with unchanged components of 1,25(OH)2D3 metabolism (i.e., CYP27B1 and CYP24A1), sodium-phosphate cotransporter 2b protein levels were not different in ileum brush borders from Uchl1-/- mice, suggesting that the intestine is not the source of hyperphosphatemia. Nonetheless, when Uchl1-/- mice were fed a low-phosphate diet, plasma phosphate, urinary phosphate, and fractional excretion of phosphate were significantly attenuated and comparable to levels of low-phosphate diet-fed wild-type mice. Our findings demonstrate that Uchl1-deleted mice exhibit perturbed phosphate homeostasis, likely consequent to decreased urinary soluble α-klotho, which can be rescued with a low-phosphate diet. Uchl1-/- mice may provide a useful mouse model to study mild perturbations in phosphate homeostasis.