Skip to Content
Merck
Haloperidol for peak identification European Pharmacopoeia (EP) Reference Standard

Haloperidol

Haloperidol for peak identification European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Haloperidol, 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone
Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique