Skip to Content
Merck
Ninhydrin spray reagent 0.2% in ethanol

Ninhydrin spray reagent

Ninhydrin spray reagent 0.2% in ethanol
Synonym(s):
Ninhydrin solution
Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14

Products

Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing