Skip to Content
Merck
Pepstatin A ≥100,000 U/mg

Pepstatin A

Pepstatin A ≥100,000 U/mg
Synonym(s):
Pepstatin A
Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
EC Number:

Products

Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing