MilliporeSigma
  • The orphan nuclear receptor Ear-2 is a negative coregulator for thyroid hormone nuclear receptor function.
The orphan nuclear receptor Ear-2 is a negative coregulator for thyroid hormone nuclear receptor function.
Molecular and cellular biology (2000-03-14)
X G Zhu, K S Park, M Kaneshige, M K Bhat, Q Zhu, C N Mariash, P McPhie, S Y Cheng
ABSTRACT
Thyroid hormone (T3) nuclear receptors (TR) are ligand-dependent transcription factors which regulate growth, differentiation, and development. One emerging hypothesis suggests that TR mediate these diverse effects via a large network of coregulators. Recently, we found that TR-mediated transcriptional responses varied in six cell lines derived from different tissues. We therefore used human TR subtype beta1 (TRbeta1) as bait to search for coregulators in human colon carcinoma RKO cells with a yeast two-hybrid system. RKO cells exhibited T3-dependent and -independent transcriptional activation. One of the three positive clones was identified as Ear-2, which is a distant member of the chick ovalbumin upstream promoter-transcription factors of the orphan nuclear receptor family. The physical interaction between Ear-2 and TRbeta1 was further confirmed by specific binding of Ear-2 to glutathione S-transferase-TRbeta1. In addition, Ear-2 was found to associate with TRbeta1 in cells. As a result of this physical interaction, binding of TRbeta1 to the T3 response elements was inhibited. Using reporter systems, we found that both the basal activation and the T3-dependent activation mediated by TRbeta1 were repressed by Ear-2 in CV1 cells. In RKO cells, however, the T3-independent transcriptional activity was more sensitive to the repression effect of Ear-2 than the T3-dependent transcriptional activity. The repression effect of Ear-2 was reversed by steroid hormone receptor coactivator 1. These results suggest that TR-mediated responses reflect a balance of corepressors and coactivators in cells. These findings further strengthen the hypothesis that the diverse activities of TR are achieved via a large network of coregulators that includes Ear-2.