MilliporeSigma
  • One-pot preparation of coenzyme A analogues via an improved chemo-enzymatic synthesis of pre-CoA thioester synthons.
One-pot preparation of coenzyme A analogues via an improved chemo-enzymatic synthesis of pre-CoA thioester synthons.
Chemical communications (Cambridge, England) (2007-01-16)
Marianne van Wyk, Erick Strauss
ABSTRACT
Coenzyme A analogues are synthesized in a one-pot preparation by biotransformation of pantothenate thioesters through the simultaneous use of three CoA biosynthetic enzymes, followed by aminolysis.
Sigma-Aldrich
5-(Biotinamido)pentylamine TFA Salt, ≥95%