MilliporeSigma
  • Promotion of Metabolite Synthesis in Isaria cicadae, a Dominant Species in the Cicada Flower Microbiota, by Cicada Pupae.
Promotion of Metabolite Synthesis in Isaria cicadae, a Dominant Species in the Cicada Flower Microbiota, by Cicada Pupae.
Journal of agricultural and food chemistry (2019-07-13)
Yan-Jie Chunyu, Zhen-Ming Lu, Zhi-Shan Luo, Shuo-Shuo Li, Hui Li, Yan Geng, Hong-Yu Xu, Zheng-Hong Xu, Jin-Song Shi
ABSTRACT
Cicada flowers, which are edible and medicinal mushrooms, are the fruiting bodies of Isaria cicadae, a fungus that is parasitic on the larvae of cicada pupae. We hypothesize that host factors might possess stimulatory activity on metabolite synthesis in Isaria cicadae. Here, we first compared the microbial community structures of different wild cicada flowers across geographical regions, compartments, and growth stages via high-throughput sequencing. Isaria cicadae TZC-3, an isolate of the most abundant operational taxonomic unit (OTU6782) in all the fungal communities, was isolated from wild cicada flowers. Furthermore, the effects of cicada pupae on metabolite synthesis in Isaria cicadae TZC-3 were studied in submerged culture. The contents of intercellular polysaccharides, adenosine, N6-(2-hydroxyethyl)-adenosine, free amino acids, and hydrolyzed monosaccharides in the mycelia cultured with cicada pupa powder (4%) were significantly increased as compared with the contents in the control group. This indicates that a cicada pupa can act as an elicitor for metabolite synthesis in Isaria cicadae.