Skip to Content
MilliporeSigma
MEH-PPV average Mn 70,000-100,000 | MEH-PPV average Mn 70,000-100,000 | 138184-36-8 average Mn 70,000-100,000

MEH-PPV average Mn 70,000-100,000 | MEH-PPV average Mn 70,000-100,000 | 138184-36-8

MEH-PPV average Mn 70,000-100,000 | MEH-PPV average Mn 70,000-100,000 | 138184-36-8 average Mn 70,000-100,000
Synonym(s):
Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], MEH-PPV
Linear Formula:
(C18H28O2)n
CAS Number:

Products

Brand
Quality Segment