WHA10342581
Whatman® Seed testing paper
L × W (420 mm) (594 mm), pack of 50 ea
About This Item
Kod UNSPSC:
41115727
NACRES:
NB.24
Polecane produkty
opakowanie
pack of 50 ea
producent / nazwa handlowa
Cytiva
Whatman 10342581
dł. × szerokość
(420 mm) (594 mm)
Powiązane kategorie
Opis ogólny
Owing to their extremely high purity Whatman seed testing papers provide reliable and reproducible results. The papers are made from pure cellulose without any additives and do not contain any substances which could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.
The better contrast of the color seed testing papers makes evaluation easier, particularly for seeds with fine white rootlets or under artificial light. This makes work easier, improves the results and saves time. The dyes which we use have been thoroughly investigated and have no influence on the growth of the seeds.
The better contrast of the color seed testing papers makes evaluation easier, particularly for seeds with fine white rootlets or under artificial light. This makes work easier, improves the results and saves time. The dyes which we use have been thoroughly investigated and have no influence on the growth of the seeds.
Cechy i korzyści
- Stała absorpcja wody przez papier zapewnia ciągłe dostarczanie wymaganej ilości wody.
Informacje prawne
Whatman is a registered trademark of Cytiva
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)