Merck
TEM-75965-01
Sigma-Aldrich
Omni Lift-Out Grid Storage Box
pkg of 1 ea
About This Item
Kod UNSPSC:
41111761
NACRES:
MA.24
opakowanie
pkg of 1 ea
Opis ogólny
Storage box for 100 standard or haft grids—3 mm diameter –TEM grids. Box comes complete with base, lid and clips.