HPLC Analysis of Metabolites of 7,12-Dimethylbenz[a]anthracene on SUPELCOSIL™ LC-18
![HPLC Analysis of Metabolites of 7,12-Dimethylbenz[a]anthracene on SUPELCOSIL™ LC-18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/268/344/9649f010-2c72-435d-be8d-ca3f1ab6272c/800/9649f010-2c72-435d-be8d-ca3f1ab6272c.jpg)
CONDITIONS
column
SUPELCOSIL LC-18, 25 cm × 4.6 mm I.D., 5 μm particles (58298)
mobile phase
[A] methanol:[B] water, (50:50, A:B), 10 min to 100:0 at 2.5% /min
flow rate
0.8 mL/min
detector
fluorescence
sample
rat liver, 9000 × g supernatant fraction from Aroclor-treated animals
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC