Merck
  • Regulation of life and death by the zinc finger transcription factor Egr-1.
Regulation of life and death by the zinc finger transcription factor Egr-1.
Journal of cellular physiology (2002-10-18)
Gerald Thiel, Giuseppe Cibelli
ABSTRACT
The biosynthesis of the zinc finger transcription factor Egr-1 is stimulated by many extracellular signaling molecules including hormones, neurotransmitters, growth and differentiation factors, and cytotoxic metabolites. The 5'-flanking region of the Egr-1 gene contains genetic elements that are essential in connecting stimulation of the cells with enhanced transcription of the Egr-1 gene, and subsequently, transcription of Egr-1-responsive genes. Thus, Egr-1 links cellular signaling cascades with changes in the gene expression pattern. Many biological functions have been attributed to Egr-1. Here, we discuss evidence for Egr-1 control of cellular proliferation and programmed cell death.