- Specificity of hemorrhagic proteinase from Crotalus atrox (western diamondback rattlesnake) venom.
Specificity of hemorrhagic proteinase from Crotalus atrox (western diamondback rattlesnake) venom.
Biochimica et biophysica acta (1985-05-20)
Y Komori, S Hagihara, A T Tu
PMID3888273
ABSTRACT
Hemorrhagic proteinase, HTb, isolated from Crotalus atrox (western diamondback rattlesnake) venom was studied for its specificity. HTb showed fibrinogenase activity, hydrolyzing the A alpha chain of fibrinogen first, followed by the cleavage of the B beta chain. HTb is different from thrombin and did not produce a fibrin clot. The degradation products of fibrinogen were found to be different, indicating that the cleavage sites in the A alpha and B beta chains are different from those of thrombin. N-Benzoyl-Phe-Val-Arg-p-nitroanilide was not hydrolyzed by HTb, although this substrate was hydrolyzed by thrombin and reptilase.