Merck
  • Iterative molecular assembly based on the cation-pool method. Convergent synthesis of dendritic molecules.
Iterative molecular assembly based on the cation-pool method. Convergent synthesis of dendritic molecules.
Journal of the American Chemical Society (2008-07-30)
Toshiki Nokami, Kousuke Ohata, Masafumi Inoue, Hiroaki Tsuyama, Akito Shibuya, Kazuya Soga, Masayuki Okajima, Seiji Suga, Jun-ichi Yoshida
ABSTRACT
An iterative method for molecular assembly has been developed based on the cation-pool method using (trimethylsilyl)diphenylmethane as a building block. The silyl group works as both an activating group of the benzene ring in the Friedel-Crafts type reaction and an electroauxiliary in the subsequent low temperature anodic oxidation to generate dendritic diarylcarbenium ions, which were well characterized by low-temperature NMR spectroscopy. The convergent synthesis of dendritic molecules has been achieved by repeating the sequence.
Sigma-Aldrich
Diphenylmethane, 99%