Merck
  • p53 and p21 expression in bone marrow clots of megaloblastic anemia patients.
p53 and p21 expression in bone marrow clots of megaloblastic anemia patients.
International journal of clinical and experimental pathology (2020-08-13)
Denis Nicolas Kwan, Júlia Thalita Queiróz Rocha, Ligia Niero-Melo, Maria Aparecida Custódio Domingues, Cristiano Claudino Oliveira
ABSTRACT
The pathogenesis of megaloblastic hemopathies (MH) is centered on the deficiency of vitamin B12 and folic acid with interruption of erythrocyte maturation. This study researched the participation of p53 and p21 in the pathophysiology of the disease. A retrospective study enrolled 95 patients with histopathologic diagnosis by biopsy or bone marrow clot (BMB/BMC), with clinical review and immunohistochemical study in tissue microarray (TMA) for p53 and p21, detailing their marking location. All patients had BMC and only 11 had BMB. The CMO was a differential of this study and it allowed an expanded sample. In the TMA, 63.7% (58/91) of the samples were immunopositive for p53; and 35.2% (31/88) were immunopositive for p21. Nuclear staining, divergent from the literature, was observed in 17.3% (10/58) among those p53+ and in 38.7% (12/31) among those p21+. The pattern of immunostaining showed non-significant differences (P=0.474) regarding morphologic and clinical aspects. The positivity for both may indicate an effective balance between apoptosis and anti-apoptotic action. Excessive inhibition of apoptosis would contribute to high global cellularity, but without functional maturation effectiveness. In conclusion, there is p21 and/or p53 immunoexpression in most cases of this study and there is no clear association between immunoexpression pattern and patient outcome. Unlike the literature, we also found a percentage of nuclear immunostaining, but the finding was not statistically significant. Combination of p21 and p53 results created different possibilities of pathologic interpretation for MH, reinforcing the importance of studies similar to this one.