Merck
  • Enantioselective reactions of donor/acceptor carbenoids derived from alpha-aryl-alpha-diazoketones.
Enantioselective reactions of donor/acceptor carbenoids derived from alpha-aryl-alpha-diazoketones.
Organic letters (2009-01-17)
Justin R Denton, Huw M L Davies
ABSTRACT
The reaction of a variety of alpha-aryl-alpha-diazo ketones with activated olefins, catalyzed by the adamantyl glycine-derived dirhodium complex Rh(2)(S-PTAD)(4), generates cyclopropyl ketones with high diastereoselectivity (up to >95:5 dr) and enantioselectivity (up to 98% ee). Intermolecular C-H functionalization of 1,4-cyclohexadiene by means of carbenoid-induced C-H insertion was also possible with this type of carbenoid.
Sigma-Aldrich
Rh2(S-PTAD)4