Merck
  • Trauma and hemorrhagic shock activate molecular association of 5-lipoxygenase and 5-lipoxygenase-Activating protein in lung tissue.
Trauma and hemorrhagic shock activate molecular association of 5-lipoxygenase and 5-lipoxygenase-Activating protein in lung tissue.
The Journal of surgical research (2018-06-26)
Geoffrey R Nunns, John R Stringham, Fabia Gamboni, Ernest E Moore, Miguel Fragoso, Gregory R Stettler, Christopher C Silliman, Anirban Banerjee
ABSTRACT
Post-traumatic lung injury following trauma and hemorrhagic shock (T/HS) is associated with significant morbidity. Leukotriene-induced inflammation has been implicated in the development of post-traumatic lung injury through a mechanism that is only partially understood. Postshock mesenteric lymph returning to the systemic circulation is rich in arachidonic acid, the substrate of 5-lipoxygenase (ALOX5). ALOX5 is the rate-limiting enzyme in leukotriene synthesis and, following T/HS, contributes to the development of lung dysfunction. ALOX5 colocalizes with its cofactor, 5-lipoxygenase-activating protein (ALOX5AP), which is thought to potentiate ALOX5 synthetic activity. We hypothesized that T/HS results in the molecular association and nuclear colocalization of ALOX5 and ALOX5AP, which ultimately increases leukotriene production and potentiates lung injury. To examine these molecular interactions, a rat T/HS model was used. Post-T/HS tissue was evaluated for lung injury through both histologic analysis of lung sections and biochemical analysis of bronchoalveolar lavage fluid. Lung tissue was immunostained for ALOX5 and ALOX5AP with association and colocalization evaluated by fluorescence resonance energy transfer. In addition, rats undergoing T/HS were treated with MK-886, a known ALOX5AP inhibitor. ALOX5 levels increase and ALOX5/ALOX5AP association occurred after T/HS, as evidenced by increases in total tissue fluorescence and fluorescence resonance energy transfer signal intensity, respectively. These findings coincided with increased leukotriene production and with the histological changes characteristic of lung injury. ALOX5/ALOX5AP complex formation, leukotriene production, and lung injury were decreased after inhibition of ALOX5AP with MK-886. These results suggest that the association of ALOX5/ALOX5AP contributes to leukotriene-induced inflammation and predisposes the T/HS animal to lung injury.