Skip to Content
Merck
SUPELCOSIL™ Suplex™ pKb-100 HPLC Column 5 μm particle size, L × I.D. 25 cm × 3 mm

SUPELCOSIL Suplex pKb-100 HPLC Column

SUPELCOSIL™ Suplex™ pKb-100 HPLC Column 5 μm particle size, L × I.D. 25 cm × 3 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique