Skip to Content
Merck
Staurosporine VETRANAL®, analytical standard

Staurosporine

Staurosporine VETRANAL®, analytical standard
Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing