Skip to Content
Merck
tert-Butylchlorodimethylsilane solution 50 wt. % in toluene

tert-Butyldimethylsilyl chloride solution

tert-Butylchlorodimethylsilane solution 50 wt. % in toluene
Synonym(s):
tert-Butylchlorodimethylsilane solution, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMSCl solution
Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2