Skip to Content
Merck
Cyanogen Bromide Activated Agarose, High Swell Ratio lyophilized powder

Cyanogen Bromide Activated Agarose, High Swell Ratio

Cyanogen Bromide Activated Agarose, High Swell Ratio lyophilized powder
Synonym(s):
Cyanogen bromide-activated Agarose
CAS Number:

Products

Brand
Biological Source
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing