Skip to Content
Merck
Agar Strips TCI-y TCI-y medium, Total count, y-irradiated, HYCON®

Agar Strips TCI-y

Agar Strips TCI-y TCI-y medium, Total count, y-irradiated, HYCON®
Synonym(s):
Agar Strips

Products

Product Category
Brand
Physical Form
Compatibility
Quality Segment
Application
Microorganism Suitability
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing