Merck
HPLC Analysis of Sugars on SUPELCOSIL LC-NH2
HPLC Analysis of Sugars on SUPELCOSIL™ LC-NH2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-NH2 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
Sucrose
suitable for microbiology, ACS reagent, ≥99.0%
Dextrose
United States Pharmacopeia (USP) Reference Standard
Fructose
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
SUPELCOSIL LC-NH2, 25 cm x 4.6 mm I.D., 5 μm particles (58338)
mobile phase
[A] water; [B] acetonitrile; (25:75, A:B)
flow rate
1 mL/min
temp.
ambient
detector
refractive index (RI)
injection
2 μL
sample
10 mg/mL in water
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC