GC Analysis of a 7-Component Sulfur Gases Mix on Supel-Q PLOT after SPME using 75 μm Carboxen/PDMS Fiber
![GC Analysis of a 7-Component Sulfur Gases Mix on Supel-Q PLOT after SPME using 75 μm Carboxen/PDMS Fiber suitable for GC, application for SPME](/deepweb/assets/sigmaaldrich/product/images/305/014/1a020807-0cae-4aa5-be3e-72cf10a32f95/800/1a020807-0cae-4aa5-be3e-72cf10a32f95.jpg)
CONDITIONS
sample/matrix
sulfur gases in 40 mL vial
SPME fiber
Carboxen/PDMS, 75 μm (57318)
extraction
10 min headspace at 40 °C
desorption process
3 min at 290 °C
column
Supel-Q PLOT, 30 m × 0.32 mm I.D. (24242)
oven
45 °C (1 min) to 250 °C at 15 °C/min
detector
FPD
Description
Legal Information
null