Merck
NFκB-Mediated Invasiveness in CD133
Molecular cancer research : MCR (2017-10-04)
Alice Nomura, Vineet K Gupta, Patricia Dauer, Nikita S Sharma, Vikas Dudeja, Nipun Merchant, Ashok K Saluja, Sulagna Banerjee
ABSTRACT
Tumor-initiating cells (TIC) have been implicated in pancreatic tumor initiation, progression, and metastasis. Among different markers that define this cell population within the tumor, the CD133