- Antibacterial Activity of 1-[(2,4-Dichlorophenethyl)amino]-3-Phenoxypropan-2-ol against Antibiotic-Resistant Strains of Diverse Bacterial Pathogens, Biofilms and in Pre-clinical Infection Models.
Antibacterial Activity of 1-[(2,4-Dichlorophenethyl)amino]-3-Phenoxypropan-2-ol against Antibiotic-Resistant Strains of Diverse Bacterial Pathogens, Biofilms and in Pre-clinical Infection Models.
Frontiers in microbiology (2018-01-10)
Valerie Defraine, Laure Verstraete, Françoise Van Bambeke, Ahalieyah Anantharajah, Eleanor M Townsend, Gordon Ramage, Romu Corbau, Arnaud Marchand, Patrick Chaltin, Maarten Fauvart, Jan Michiels
PMID29312259
ABSTRACT
We recently described the novel anti-persister compound 1-[(2,4-dichlorophenethyl)amino]-3-phenoxypropan-2-ol (SPI009), capable of directly killing persister cells of the Gram-negative pathogen