Merck
CCDC88B is required for pathogenesis of inflammatory bowel disease.
Nature communications (2017-10-17)
Nassima Fodil, Neda Moradin, Vicki Leung, Jean-Frederic Olivier, Irena Radovanovic, Thiviya Jeyakumar, Manuel Flores Molina, Ashley McFarquhar, Romain Cayrol, Dominique Bozec, Naglaa H Shoukry, Michiaki Kubo, Julia Dimitrieva, Edouard Louis, Emilie Theatre, Stephanie Dahan, Yukihide Momozawa, Michel Georges, Garabet Yeretssian, Philippe Gros
ABSTRACT
Inflammatory bowel disease (IBD) involves interaction between host genetic factors and environmental triggers. CCDC88B maps within one IBD risk locus on human chromosome 11q13. Here we show that CCDC88B protein increases in the colon during intestinal injury, concomitant with an influx of CCDC88B