Merck
  • Downregulation of esophageal cancer-related gene 4 promotes proliferation and migration of hepatocellular carcinoma.
Downregulation of esophageal cancer-related gene 4 promotes proliferation and migration of hepatocellular carcinoma.
Oncology letters (2017-09-21)
Shujian Ge, Yali Xu, Hongliang Wang, Yaxin Sun, Xiangguo Tian, Zhixin Cao, Xiaoyan Lin, Jiawen Xu, Qiangxiu Wang
ABSTRACT
Esophageal cancer-related gene 4 (ECRG4) is a candidate tumor suppressor gene, which is involved in cell apoptosis, migration, infection and inflammation responsiveness; however, its expression level and clinical significance in hepatocellular carcinoma (HCC) remains unclear. In the present study, the authors aim to evaluate the clinical significance and potential role of ECRG4 in HCC. Level of ECRG4 protein expression in HCC and peripheral tissues was investigated in tissue specimens obtained from 56 consecutive HCC patients by immunohistochemistry. Cell proliferation, cell migration and invasion regulations were examined by MTT curves, flow cytometry, Transwell assays and western blotting. ECRG4 expression was weak positive in normal liver cells but was downregulated in HCC cells