Merck
  • Suppression of Npr1, not Npr2 gene function induces hypertrophic growth in H9c2 cells in vitro.
Suppression of Npr1, not Npr2 gene function induces hypertrophic growth in H9c2 cells in vitro.
Biochemical and biophysical research communications (2017-07-27)
Senthamizharasi Manivasagam, Elangovan Vellaichamy
ABSTRACT
Npr1 gene (coding for NPR-A) and Npr2 gene (coding for NPR-B) are identified as intrinsic anti-hypertrophic genes that opposes abnormal cardiac remodeling. However, the functional role of Npr1 and Npr2 genes during cardiac hypertrophic growth is not well understood. Hence, the present investigation was aimed to study the effect of Npr1 and Npr2 gene silencing, respectively on β-AR activation induced cardiac hypertrophic growth in H9c2 cells in vitro. The control, Npr1, and Npr2 gene suppressed H9c2 cells, respectively were treated with ISO (10