- Localization patterns of the ganglioside G
Localization patterns of the ganglioside G
Molecular reproduction and development (2017-04-19)
Cristina Cardona, Queenie V Neri, Alana J Simpson, Melissa A Moody, G Charles Ostermeier, Eric K Seaman, Theodore Paniza, Zev Rosenwaks, Gianpiero D Palermo, Alexander J Travis
PMID28418610
ABSTRACT
Semen analysis lacks a functional component and best identifies extreme cases of infertility. The ganglioside G