Merck
  • Cord blood DNA methylation and adiposity measures in early and mid-childhood.
Cord blood DNA methylation and adiposity measures in early and mid-childhood.
Clinical epigenetics (2017-08-18)
Jacob K Kresovich, Yinan Zheng, Andres Cardenas, Brian T Joyce, Sheryl L Rifas-Shiman, Emily Oken, Matthew W Gillman, Marie-France Hivert, Andrea A Baccarelli, Lifang Hou
ABSTRACT
Excess adiposity in childhood is associated with numerous adverse health outcomes. As this condition is difficult to treat once present, identification of risk early in life can help inform and implement strategies to prevent the onset of the condition. We performed an epigenome-wide association study to prospectively investigate the relationship between cord blood DNA methylation and adiposity measurements in childhood. We measured genome-wide DNA methylation from 478 children in cord blood and measured overall and central adiposity via skinfold caliper measurements in early (range 3.1-3.3 years) and mid-childhood (age range 7.3-8.3 years) and via dual X-ray absorptiometry (DXA) in mid-childhood. Final models were adjusted for maternal age at enrollment, pre-pregnancy body mass index, education, folate intake during pregnancy, smoking during pregnancy, and gestational weight gain, and child sex, race/ethnicity, current age, and cord blood cell composition. We identified four promoter proximal CpG sites that were associated with adiposity as measured by subscapular (SS) and triceps (TR) ratio (SS:TR) in early childhood, in the genes This analysis identified novel CpG loci associated with adiposity outcomes. However, our results suggest little consistency across the various adiposity outcomes tested, particularly among the more accurate DXA measurements of body composition. We recommend using caution when interpreting these associations.